siRNA Details
Gene Symbol: 4930511M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015494.1 | 2, 3 | 438 | |||
NR_027945.1 | 2, 3 | 268 | ||||
GenBank mRNA | AK015760.1 | - | 2, 3 | 281 | ||
AK144587.1 | - | 2, 3 | 406 | |||
AK165876.1 | - | 2, 3 | 439 |
