siRNA Details
Gene Symbol: BC046251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149279.1 | - | 4 | 2363 | ||
BC046251.1 | - | 6 | 955 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC046251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149279.1 | - | 4 | 2363 | ||
BC046251.1 | - | 6 | 955 |