siRNA Details
Gene Symbol: AI427809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033139.1 | 2 | 913 | |||
NR_033140.1 | 2 | 1003 | ||||
GenBank mRNA | AK144542.1 | - | 2 | 915 | ||
AK171862.1 | - | 2 | 932 | |||
BC086687.1 | - | 2 | 1003 | |||
BC147516.1 | - | 1 | 72 | 9056508 | ||
BC147547.1 | - | 1 | 72 | 9056538 |
