siRNA Details
Gene Symbol: Gm15997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045423.1 | 3, 4 | 436 | |||
GenBank mRNA | AK043895.1 | - | 3, 4 | 437 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB641762.1 | - | 3, 4 | 437 | Chr. 5 - 149493165 - 149538030 [-] on Build GRCm38 |
