siRNA Details
Gene Symbol: 4930512H18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI037313.1 | - | 2 | 138 | ||
AK015772.1 | - | 2 | 174 | |||
AK084432.1 | - | 282 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB658464.1 | - | 3 | 282 | Chr. 8 - 61849006 - 61921991 [+] on Build GRCm38 |
