siRNA Details
Gene Symbol: 4933431E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015459.1 | 3 | 2145 | |||
GenBank mRNA | AK030251.1 | - | 4 | 1768 | ||
AK036227.1 | - | 3 | 1718 | |||
AK147824.1 | - | 4 | 1742 | |||
BC060136.1 | - | 3 | 2145 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177858.1 | - | 1718 | Chr. - - [-] on Build |
