siRNA Details
Gene Symbol: 4930442L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015596.1 | 1, 2 | 56 | |||
GenBank mRNA | AK006795.1 | - | 1 | 231 | ||
AK006978.1 | - | 1, 2 | 36 | |||
AK015364.1 | - | 1, 2 | 56 |
