siRNA Details
Gene Symbol: 2610035D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015556.1 | 3 | 618 | |||
GenBank mRNA | AK011682.1 | - | 3 | 161 | ||
AK031204.1 | - | 3 | 619 | |||
BC024713.1 | - | 2 | 557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610035D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015556.1 | 3 | 618 | |||
GenBank mRNA | AK011682.1 | - | 3 | 161 | ||
AK031204.1 | - | 3 | 619 | |||
BC024713.1 | - | 2 | 557 |