siRNA Details
Gene Symbol: Gm16725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040746.1 | - | 4 | 1160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB214750.2 | - | 3 | 626 | Chr. 8 - 18652801 - 18676419 [-] on Build GRCm38 |
