siRNA Details
Gene Symbol: C430049B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012483.1 | - | 2 | 750 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711610.1 | - | 2 | 745 | Chr. X - 53056193 - 53057188 [-] on Build GRCm38 |
