siRNA Details
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040257.1 | 3 | 517 | |||
NR_040258.1 | 2 | 263 | ||||
NR_040259.1 | 2 | 263 | ||||
NR_040261.1 | 2 | 263 | ||||
NR_040262.1 | 2 | 263 | ||||
GenBank mRNA | AA427326.1 | - | 2 | 188 | ||
AW105955.1 | - | 2 | 171 | |||
BC021953.1 | - | 3 | 424 | |||
BC099512.1 | - | 2 | 183 | |||
BY261359.1 | - | 2 | 264 |
