siRNA Details
Gene Symbol: 2310065F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038055.1 | 4 | 824 | |||
GenBank mRNA | AK010044.1 | - | 4 | 824 | ||
AK079404.1 | - | 3 | 825 | |||
AK080188.1 | - | 2 | 531 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310065F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038055.1 | 4 | 824 | |||
GenBank mRNA | AK010044.1 | - | 4 | 824 | ||
AK079404.1 | - | 3 | 825 | |||
AK080188.1 | - | 2 | 531 |