siRNA Details
Gene Symbol: 5730405O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038158.1 | 3 | 544 | |||
GenBank mRNA | AK017500.1 | - | 3 | 509 | ||
AK079682.1 | - | 1 | 555 | |||
AK134096.1 | - | 2 | 1493 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717287.1 | - | 3 | 503 | Chr. X - 13042056 - 13045295 [+] on Build GRCm38 |
