siRNA Details
Gene Symbol: E130018N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040327.1 | 2 | 427 | |||
GenBank mRNA | AK018699.1 | - | 2 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY731227.1 | - | 2 | 390 | Chr. 2 - 168152927 - 168154476 [-] on Build GRCm38 |
