siRNA Details
Gene Symbol: B230216N24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037993.1 | 3 | 448 | |||
GenBank mRNA | AK021003.1 | - | 3 | 415 | ||
BC049988.1 | - | 2 | 2732 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727151.1 | - | 3 | 415 | Chr. 1 - 98031586 - 98047759 [-] on Build GRCm38 |
