siRNA Details
Gene Symbol: Gm15759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028685.1 | - | 612 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB612936.1 | - | 3 | 610 | Chr. 1 - 58289429 - 58315655 [-] on Build GRCm38 |
