siRNA Details
Gene Symbol: 4930428N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019699.1 | - | 2 | 317 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715452.1 | - | 2 | 317 | Chr. 14 - 24622138 - 24622862 [+] on Build GRCm38 |
