siRNA Details
Gene Symbol: A430072P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138808.1 | - | 1 | 185 | ||
AK146255.1 | - | 2 | 734 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A430072P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138808.1 | - | 1 | 185 | ||
AK146255.1 | - | 2 | 734 |