siRNA Details
Gene Symbol: 2810442N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040562.1 | 3 | 837 | |||
GenBank mRNA | AK013293.1 | - | 3 | 838 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2810442N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040562.1 | 3 | 837 | |||
GenBank mRNA | AK013293.1 | - | 3 | 838 |