siRNA Details
Gene Symbol: 4930518C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015822.1 | - | 2 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715366.1 | - | 2 | 284 | Chr. 5 - 48003066 - 48005704 [-] on Build GRCm38 |
