siRNA Details
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015385.2 | 3 | 636 | |||
NR_015386.2 | 3 | 707 | ||||
NR_038082.1 | 4 | 2748 | ||||
NR_038083.1 | 4 | 1832 | ||||
NR_038084.1 | 4 | 625 | ||||
GenBank mRNA | AK053722.1 | - | 4 | 1834 | ||
AY589790.1 | - | 3 | 586 | |||
AY590888.1 | - | 4 | 454 | |||
AY590889.1 | - | 3 | 464 | |||
BC065087.1 | - | 3 | 3632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE949511.1 | - | 3 | 586 | Chr. 17 - 85611545 - 85618275 [-] on Build GRCm38 |
