siRNA Details
Gene Symbol: Gm20611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040638.1 | 3 | 964 | |||
GenBank mRNA | AK029795.1 | - | 3 | 964 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB016902.2 | - | 2 | 523 | Chr. 10 - 61036723 - 61038080 [+] on Build GRCm38 |
