siRNA Details
Gene Symbol: B130034C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040375.1 | 3 | 817 | |||
GenBank mRNA | AK045112.1 | - | 3 | 818 | ||
AK139053.1 | - | 2 | 1756 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B130034C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040375.1 | 3 | 817 | |||
GenBank mRNA | AK045112.1 | - | 3 | 818 | ||
AK139053.1 | - | 2 | 1756 |