siRNA Details
Gene Symbol: 1700055C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040726.1 | 3 | 233 | |||
GenBank mRNA | AK006792.1 | - | 3 | 234 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV204450.1 | - | 3 | 180 | Chr. 9 - 64038465 - 64041406 [-] on Build GRCm38 |
