siRNA Details
Gene Symbol: 4930535L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019721.1 | - | 3 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715554.1 | - | 3 | 249 | Chr. 9 - 107998116 - 108002404 [-] on Build GRCm38 |
