siRNA Details
Gene Symbol: Snhg18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038186.1 | 2 | 1114 | |||
GenBank mRNA | AK002304.1 | - | 2 | 1114 | ||
AK007520.1 | - | 2 | 873 | |||
BX528012.1 | - | 1 | 500 | |||
CJ321181.1 | - | 1 | 80 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU509963.1 | - | 2 | 890 | Chr. 15 - 32240674 - 32244411 [-] on Build GRCm38 |
