siRNA Details
Gene Symbol: Gm26710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133627.1 | - | 495 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ048224.1 | - | 2 | 495 | Chr. 10 - 80578647 - 80579496 [+] on Build GRCm38 |
