siRNA Details
Gene Symbol: Pvt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003368.1 | 6 | 921 | |||
GenBank mRNA | AK020628.1 | - | 4 | 652 | ||
AK043252.1 | - | 307 | ||||
AK043790.1 | - | 5 | 776 | |||
AK090048.1 | - | 644 | ||||
AK169445.1 | - | 6 | 862 | |||
BC049751.1 | - | 2 | 169 | |||
Z12002.1 | - | 2 | 161 |
