siRNA Details
Gene Symbol: 4933440J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045344.1 | 3 | 248 | |||
GenBank mRNA | AK017133.1 | - | 3 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716812.1 | - | 3 | 249 | Chr. 10 - 111586917 - 111594273 [-] on Build GRCm38 |
