siRNA Details
Gene Symbol: 1810034E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045798.1 | 2, 3 | 686 | |||
GenBank mRNA | AK002708.1 | - | 2, 3 | 334 | ||
AK007690.1 | - | 2, 3 | 686 | |||
AK143964.1 | - | 2, 3 | 641 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703213.1 | - | 2, 3 | 334 | Chr. 13 - 64249052 - 64267496 [+] on Build GRCm38 |
