siRNA Details
Gene Symbol: Gm4890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045822.1 | 3 | 1802 | |||
NR_045823.1 | 2 | 1762 | ||||
GenBank mRNA | AK041972.1 | - | 3 | 1758 | ||
AK137125.1 | - | 1060 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045822.1 | 3 | 1802 | |||
NR_045823.1 | 2 | 1762 | ||||
GenBank mRNA | AK041972.1 | - | 3 | 1758 | ||
AK137125.1 | - | 1060 |