siRNA Details
Gene Symbol: 2310002F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077063.1 | 2 | 217 | |||
GenBank mRNA | AK009091.1 | - | 2 | 218 | ||
AK134755.1 | - | 1005 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708938.1 | - | 2 | 218 | Chr. 7 - 43747189 - 43755936 [-] on Build GRCm38 |
