siRNA Details
Gene Symbol: A330076H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015599.2 | 1 | 291 | |||
GenBank mRNA | AK035901.1 | - | 1 | 345 | ||
AK036297.1 | - | 1 | 292 | |||
AK038438.1 | - | 345 | ||||
AK039637.1 | - | 1 | 319 | |||
AK080655.2 | - | 348 | ||||
AK083195.1 | - | 1 | 344 |
