siRNA Details
Gene Symbol: Gm16907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045794.1 | 3 | 1801 | |||
GenBank mRNA | AK030196.1 | - | 3 | 1802 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045794.1 | 3 | 1801 | |||
GenBank mRNA | AK030196.1 | - | 3 | 1802 |