siRNA Details
Gene Symbol: Gm6634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040556.1 | 4 | 631 | |||
GenBank mRNA | AK144724.1 | - | 4 | 632 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734237.1 | - | 4 | 631 | Chr. 3 - 70773527 - 70807291 [-] on Build GRCm38 |
