siRNA Details
Gene Symbol: 1700034B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006590.1 | - | 2 | 461 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706592.1 | - | 2 | 461 | Chr. 18 - 75781031 - 75782303 [+] on Build GRCm38 |
