siRNA Details
Gene Symbol: Rmst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028262.1 | 2 | 100 | |||
GenBank mRNA | AK049726.1 | - | 2 | 65 | ||
AK082467.1 | - | 2 | 93 | |||
AK086758.1 | - | 2 | 93 | |||
BB621570.1 | - | 2 | 101 |
