siRNA Details
Gene Symbol: D930016D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030673.1 | 4 | 1342 | |||
GenBank mRNA | AK040297.1 | - | 3 | 1703 | ||
AK086246.1 | - | 1 | 1101 | |||
AK140900.1 | - | 2 | 897 | |||
BC060616.1 | - | 4 | 1309 | |||
BC082597.1 | - | 1818 |
