siRNA Details
Gene Symbol: Gm26744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138627.1 | - | 1 | 123 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724381.1 | - | 1 | 123 | Chr. 16 - 21844482 - 21845776 [-] on Build GRCm38 |
