siRNA Details
Gene Symbol: Gm6277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045421.1 | 4 | 692 | |||
GenBank mRNA | AK140316.1 | - | 4 | 694 | ||
BC030050.1 | - | 4 | 652 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045421.1 | 4 | 692 | |||
GenBank mRNA | AK140316.1 | - | 4 | 694 | ||
BC030050.1 | - | 4 | 652 |