siRNA Details
Gene Symbol: D430036J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040393.1 | 5 | 780 | |||
NR_040394.1 | 4 | 757 | ||||
NR_040395.1 | 3 | 654 | ||||
NR_040396.1 | 5 | 553 | ||||
NR_040397.1 | 4 | 530 | ||||
GenBank mRNA | AK052495.1 | - | 5 | 780 | ||
AK082555.1 | - | 3 | 1034 | |||
AK162636.1 | - | 4 | 532 | |||
BB635290.1 | - | 5 | 493 |
