siRNA Details
Gene Symbol: D430036J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040393.1 | 4 | 563 | |||
NR_040394.1 | 3 | 540 | ||||
NR_040395.1 | 2 | 437 | ||||
NR_040396.1 | 4 | 336 | ||||
NR_040397.1 | 3 | 313 | ||||
GenBank mRNA | AK052495.1 | - | 4 | 563 | ||
AK082555.1 | - | 2 | 817 | |||
AK162636.1 | - | 3 | 315 | |||
BB635290.1 | - | 4 | 276 | |||
CX242024.1 | - | 2 | 420 |
