siRNA Details
Gene Symbol: 2610020C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038156.1 | 3 | 955 | |||
GenBank mRNA | AK029005.1 | - | 2 | 1726 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610020C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038156.1 | 3 | 955 | |||
GenBank mRNA | AK029005.1 | - | 2 | 1726 |