siRNA Details
Gene Symbol: 4930477E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015579.1 | - | 2 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715128.1 | - | 2 | 382 | Chr. 3 - 96401167 - 96402014 [-] on Build GRCm38 |
