siRNA Details
Gene Symbol: Gm26562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132624.1 | - | 2 | 507 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714286.1 | - | 2 | 507 | Chr. 16 - 30202285 - 30205879 [-] on Build GRCm38 |
