siRNA Details
Gene Symbol: Gm26608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040735.1 | - | 1133 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB214622.2 | - | 1 | 427 | Chr. 5 - 28053951 - 28054608 [-] on Build GRCm38 |
