siRNA Details
Gene Symbol: 4930471M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045980.1 | 1 | 392 | |||
GenBank mRNA | AK015209.1 | - | 1 | 451 | ||
AK015554.1 | - | 1 | 393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715104.1 | - | 1 | 393 | Chr. 6 - 91430616 - 91440854 [-] on Build GRCm38 |
