siRNA Details
Gene Symbol: 4931440J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045503.1 | 6, 7 | 535 | |||
GenBank mRNA | AK016511.1 | - | 6, 7 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716190.1 | - | 6, 7 | 533 | Chr. 14 - 62760503 - 62830127 [+] on Build GRCm38 |
