siRNA Details
Gene Symbol: Gm16794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040392.1 | - | 282 | |||
AK053748.1 | - | 889 | ||||
AK166469.1 | - | 2 | 478 | |||
AK169482.1 | - | 2 | 488 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040392.1 | - | 282 | |||
AK053748.1 | - | 889 | ||||
AK166469.1 | - | 2 | 478 | |||
AK169482.1 | - | 2 | 488 |