siRNA Details
Gene Symbol: 9630001P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102378.1 | 3 | 469 | |||
GenBank mRNA | AK035757.1 | - | 977 | |||
AK044526.1 | - | 3 | 329 | |||
AK080411.1 | - | 3 | 457 | |||
BU610152.1 | - | 3 | 469 |
